Learn More
2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g
Description
2-Chlorotrityl chloride is used as an intermediate of clotrimazole. It is also used in convergent peptide synthesis. Furthermore, it is used in the preparation of modified trityl nucleosides. In addition, it serves as a protecting group reagent for carboxylic acids forming esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Physical Form | Powder |
| Quantity | 10 g |
| Mesh Size | 100 to 200 Mesh |
| Chemical Name or Material | 2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 1.0 to 1.4 mmol/g |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.