missing translation for 'onlineSavingsMsg'
Learn More

2-Coumaranone, 97%

Product Code. 10514091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10514091 5 g 5g
10071760 1 g 1g
10256200 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10514091 Supplier Thermo Scientific Acros Supplier No. 154460050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 553-86-6 | C8H6O2 | 134.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-86-6
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00005856
InChI Key ACZGCWSMSTYWDQ-UHFFFAOYSA-N
Synonym 2-coumaranone, benzofuran-2 3h-one, 2 3h-benzofuranone, 3h-benzofuran-2-one, benzofuran-2-one, benzofuranone, 1-benzofuran-2 3h-one, isocoumaranone, 2-coumarotioiie
PubChem CID 68382
IUPAC Name 3H-1-benzofuran-2-one
SMILES C1C2=CC=CC=C2OC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 47°C to 52°C
Boiling Point 248°C to 250°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 309
Formula Weight 134.13
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Coumaranone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Coumaranone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.