missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyano-3-chloropyridine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-Cyano-3-chloropyridine |
| CAS | 38180-46-0 |
| Molecular Formula | C6 H3 Cl N2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C6H3ClN2/c7-5-2-1-3-9-6(5)4-8/h1-3H |
| SMILES | Clc1cccnc1C#N |
| IUPAC Name | 3-chloropyridine-2-carbonitrile |
| Molecular Weight (g/mol) | 138.55 |
| Formula Weight | 137.9985 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction