Learn More
2-Cyano-3-iodopyridine, 95%
Description
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
Specifications
| Melting Point | 98°C to 100°C |
| Quantity | 1 g |
| UN Number | UN3439 |
| Sensitivity | Light sensitive |
| Formula Weight | 230.01 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-Cyano-3-iodopyridine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.