missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-iodopyridine, 95%

Product Code. 15443577
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443577 250 mg 250mg
11491570 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443577 Supplier Thermo Scientific Alfa Aesar Supplier No. H26963.MD
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 827616-52-4 | C6H3IN2 | 230.008 g/mol

Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827616-52-4
Molecular Formula C6H3IN2
Molecular Weight (g/mol) 230.008
MDL Number MFCD09037451
InChI Key MZZGCCSFZCRVAF-UHFFFAOYSA-N
Synonym 3-iodopicolinonitrile, 2-cyano-3-iodopyridine, 2-pyridinecarbonitrile,3-iodo, pubchem15743
PubChem CID 12180890
IUPAC Name 3-iodopyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)I

Specifications

Melting Point 98°C to 100°C
Quantity 250 mg
UN Number UN3439
Sensitivity Light sensitive
Formula Weight 230.01
Percent Purity 95%
Chemical Name or Material 2-Cyano-3-iodopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.