missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-methoxybenzeneboronic acid, 95%

Product Code. 15488438
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488438 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15488438 Supplier Thermo Scientific Alfa Aesar Supplier No. H61856.MC
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1164100-84-8 | C8H8BNO3 | 176.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1164100-84-8
Molecular Formula C8H8BNO3
MDL Number MFCD10696671
Quantity 100 mg
Synonym 2-cyano-3-methoxyphenyl boronic acid, acmc-2099sp, 2-cyano-3-methoxyphenyl boronicacid
InChI Key GRHINHWGIHWEPE-UHFFFAOYSA-N
SMILES COC1=CC=CC(B(O)O)=C1C#N
IUPAC Name (2-cyano-3-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 176.97
PubChem CID 53216734
Formula Weight 176.97
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Cyano-3-methoxybenzeneboronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.