missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10482213
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10482213 250 mg Amber Glass Bottle 250mg
10202842 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10482213 Supplier Thermo Scientific Maybridge Supplier No. MO00753CB
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 197960-31-9
Molecular Formula C8H6ClNO2S
Molecular Weight (g/mol) 215.65
MDL Number MFCD03659720
InChI Key ZFQDREVAYVNITB-UHFFFAOYSA-N
Synonym benzenesulfonylchloride, 2-cyano-5-methyl, 2-cyano-5-methyl-benzenesulfonyl chloride, 2-cyano-5-methylbenzene-1-sulfonyl chloride, 3-chlorosulphonyl-4-cyanotoluene, 2-cyano-5-methylbenzenesulfonylchloride, 2-chlorosulphonyl-4-methylbenzonitrile, 2-cyano-5-methylbenzene-sulfonyl chloride, 2-cyano-5-methylbenzenesulphonyl chloride, 2-cyano-5-methylphenylsulfonyl chloride, 2-chlorosulfonyl-4-methylbenzenecarbonitrile
PubChem CID 2779837
SMILES CC1=CC(=C(C=C1)C#N)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2-cyano-5-methylbenzenesulfonyl chloride
Formula Weight 215.66
Percent Purity 97%
Chemical Name or Material 2-Cyano-5-methylbenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.