missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-(trifluoromethyl)pyridine, 98%

Product Code. 15451719
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451719 5 g 5g
15461719 250 mg 250mg
15441719 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451719 Supplier Thermo Scientific Alfa Aesar Supplier No. H64486.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95727-86-9 | C7H3F3N2 | 172.11 g/mol

It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95727-86-9
Molecular Formula C7H3F3N2
Molecular Weight (g/mol) 172.11
MDL Number MFCD01001119
InChI Key WDSCJULUXJSJOX-UHFFFAOYSA-N
Synonym 5-trifluoromethyl picolinonitrile, 5-trifluoromethyl pyridine-2-carbonitrile, 2-cyano-5-trifluoromethyl pyridine, 2-cyano-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinecarbonitrile, 5-trifluoromethyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 5-trifluoromethyl, pubchem17518, 5-trifluoromethyl ;picolinonitrile, 2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile
PubChem CID 1535372
IUPAC Name 5-(trifluoromethyl)pyridine-2-carbonitrile
SMILES FC(F)(F)C1=CN=C(C=C1)C#N

Specifications

Quantity 5 g
UN Number UN3439
Beilstein 13339826
Solubility Information Slightly soluble in water.
Formula Weight 172.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Cyano-5-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.