missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoacetamide, 99%

Product Code. p-94998 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10001560

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-91-5 | C3H4N2O | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-91-5
Molecular Formula C3H4N2O
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008024
InChI Key DGJMPUGMZIKDRO-UHFFFAOYSA-N
Synonym cyanoacetamide, acetamide, 2-cyano, cyanacetamide, malonamide nitrile, malonamonitrile, nitrilomalonamide, cyanoiminoacetic acid, 3-nitrilo-propionamide, propionamide, 3-nitrilo, usaf kf-14
PubChem CID 7898
IUPAC Name 2-cyanoacetamide
SMILES NC(=O)CC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 118°C to 122°C
Color White to Cream
Flash Point 215°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula NCCH2CONH2
Quantity 100 g
Beilstein 02, 589
Merck Index 15, 2678
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 84.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanoacetamide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyanoacetamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.