missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzeneboronic acid pinacol ester, 98%

Product Code. 11387339
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387339 5 g 5g
11397339 1 g 1g
11307349 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387339 Supplier Thermo Scientific Alfa Aesar Supplier No. H27469.06
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 214360-48-2 | C13H16BNO2 | 229.086 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214360-48-2
Molecular Formula C13H16BNO2
Molecular Weight (g/mol) 229.086
MDL Number MFCD04038747
InChI Key SKQNWSBNAIOCOC-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyanophenylboronic acid pinacol ester, 2-cyanophenylboronic acid, pinacol ester, 2-cyanobenzeneboronic acid pinacol ester, benzonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-2-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, pubchem7921, ksc925c3p
PubChem CID 3570229
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C#N

Specifications

Melting Point 88°C to 90°C
Quantity 5 g
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Cyanobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.