missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethylhydrazine, 95%, Thermo Scientific Chemicals

Product Code. 11430748
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430748 25 g 25g
11410748 1 g 1g
11420748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430748 Supplier Thermo Scientific Alfa Aesar Supplier No. B25227.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanoethylhydrazine is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-07-1
Molecular Formula C3H7N3
Molecular Weight (g/mol) 85.11
MDL Number MFCD00043620
InChI Key VIHRIIARIFUQLC-UHFFFAOYSA-N
Synonym 2-cyanoethylhydrazine, beta-cyanoethylhydrazine, 2-cyanoethyl hydrazine, cyanoethylhydrazine, unii-l6h109c98b, 3-hydrazinopropionitrile, propanenitrile, 3-hydrazino, 3-hydrazinopropanenitrile
PubChem CID 136177
IUPAC Name 3-hydrazinylpropanenitrile
SMILES C(CNN)C#N

Specifications

Density 1.053
Boiling Point 76°C to 79°C (0.5 mmHg)
Flash Point 109°C (228°F)
Linear Formula NCCH2CH2NHNH2
Refractive Index 1.477
Quantity 25 g
UN Number UN2927
Beilstein 773834
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 85.11
Percent Purity 95%
Chemical Name or Material 2-Cyanoethylhydrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.