missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanopyridine-5-boronic acid pinacol ester, 96%

Product Code. 15471486
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471486 250 mg 250mg
15440908 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471486 Supplier Thermo Scientific Alfa Aesar Supplier No. H54689.MD
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 741709-63-7 | C12H15BN2O2 | 230.07 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is used in suzuki reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is used in suzuki reactions.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents, air, heat. Refrigerated (2-4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 741709-63-7
Molecular Formula C12H15BN2O2
Molecular Weight (g/mol) 230.07
MDL Number MFCD06657825
InChI Key IXTBQKLZPOYJFJ-UHFFFAOYSA-N
Synonym 2-cyanopyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carbonitrile, 2-cyano pyridine-5-boronic acid pinacol ester, 2-cyanopyridin-5-yl boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carbonitrile, 2-cyanopyridine-5-boronic acid pinacole ester, 6-cyanopyridine-3-boronic acid pinacol ester, 2-pyridinecarbonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarbonitrile
PubChem CID 16414188
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C#N

Specifications

Melting Point 105°C to 106°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 230.07
Percent Purity 96%
Chemical Name or Material 2-Cyanopyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.