missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanopyridine, 99%

Product Code. 10142280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10142280 5 g Glass bottle 5g
10419020 100 g Glass bottle 100g
10356220 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10142280 Supplier Thermo Scientific Acros Supplier No. 110850050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-70-9 | C6H4N2 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-70-9
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00006218
InChI Key FFNVQNRYTPFDDP-UHFFFAOYSA-N
Synonym 2-cyanopyridine, picolinonitrile, 2-pyridinecarbonitrile, pyridinecarbonitrile, 2-pyridyl nitrile, picolinic acid nitrile, cyanopyridine, unii-whr1dpg7ys, 2-pyridinecarboxylic acid, nitrile, 2-cyano pyridine
PubChem CID 7522
ChEBI CHEBI:27837
IUPAC Name pyridine-2-carbonitrile
SMILES C1=CC=NC(=C1)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 26.0°C to 28.0°C
Color White to Brown
Boiling Point 212.0°C to 215.0°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CN
Refractive Index 1.5288
Quantity 5 g
Beilstein 22, 36
Solubility Information Solubility in water: immiscible.
Formula Weight 104.11
Percent Purity 99%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material 2-Cyanopyridine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyanopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.