missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethylboronic acid pinacol ester, 96%

Product Code. 15424047
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424047 1 g 1g
15434047 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424047 Supplier Thermo Scientific Alfa Aesar Supplier No. H31156.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 167692-95-7 | C14H27BO2 | 238.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167692-95-7
Molecular Formula C14H27BO2
Molecular Weight (g/mol) 238.178
MDL Number MFCD12546193
InChI Key LDKDGYZAIIKCGK-UHFFFAOYSA-N
Synonym 2-cyclohexylethylboronic acid pinacol ester, 2-2-cyclohexylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-cyclohexylethylboronicacid
PubChem CID 15259990
IUPAC Name 2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2CCCCC2

Specifications

Quantity 1 g
Formula Weight 238.18
Percent Purity 96%
Chemical Name or Material 2-Cyclohexylethylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.