missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyclopenten-1-one, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Cyclopenten-1-one |
| CAS | 930-30-3 |
| Molecular Formula | C5H6O |
| Quantity | 10 g |
| Synonym | 1-Cyclopenten-3-one, 1-Cyclopenten-5-one, 2-Cyclopentenone, Cyclopent-2-en-1-one, Cyclopenten-3-one, NSC 73117 |
| InChI Formula | InChI=1S/C5H6O/c6-5-3-1-2-4-5/h1,3H,2,4H2 |
| SMILES | O=C1CCC=C1 |
| IUPAC Name | cyclopent-2-en-1-one |
| Molecular Weight (g/mol) | 82.1 |
| Formula Weight | 82.0419 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction