missing translation for 'onlineSavingsMsg'
Learn More

Cytiva 2-D Clean-up Kit

Product Code. 10298894 Shop All Cytiva Products
Change view
Click to view available options
Sufficient For:
50 samples of less than 100μL
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
10298894 50 samples of less than 100μL 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10298894 Supplier Cytiva Supplier No. 80648451
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Improve the quality of 2-D electrophoresis. Cytiva Life Sciences™ 2-D Clean-up Kit removes interfering contaminants for 2-D electrophoresis.

  • Quantitative precipitation separates proteins from detergents, salts, lipids, phenolics and nucleic acids
  • Does not result in spot gain or loss
  • Processes 50 samples of up to 100μL each, but is scaleable for larger volumes or more diliute samples
  • Delivers yield in about 90 minutes

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1600°C
Color Colorless
Form Liquid
For Use With (Application) Improves quality of 2-D electrophoresis
Kit Contents 15mL Precipitant, 17.5mL coprecipitant, 50mL wash Buffer, 250μL wash additive, quick reference card, instructions
Melting Temperature 851°C
Molecular Weight (g/mol) 105.98874
Storage Requirements −20°C
Sufficient For 50 samples of less than 100μL
Product Type Electrophoresis Clean-up Kit
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-D Clean-Up Kit, 50 samples
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling equipment/ equipment.
  • P331-Do NOT induce vomiting.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305-IF IN EYES:.
  • P235-Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium carbonate, Cholan-24-oic acid, 3,12-dihydroxy-, monosodium salt, monohydrate, (3.alpha., 5.b
  • UNKNOWN ACUTE-5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 9.9% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.