missing translation for 'onlineSavingsMsg'
Learn More

Cytiva 2-D Quant Kit

Product Code. 10094634 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 Assays
Unit Size:
1kg
Product Code. Quantity unitSize
10094634 500 Assays 1kg
1 options
This item is not returnable. View return policy

Product Code. 10094634

Brand: Cytiva 80648356

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Precipitant, 250mL; Co-precipitant, 250mL; Copper solution, 50mL; Color reagent A, 2 x 250mL; Color reagent B, 5mL; BSA standard (2mg/mL), 5mL

Use for accurate determination of protein concentration in samples prepared for electrophoresis techniques. Cytiva Life Sciences™ 2-D Quant Kit precipitates proteins while leaving interfering substances behind.

Designed for the accurate determination of protein concentration in samples prepared for electrophoresis techniques. Works by quantitatively precipitating proteins while leaving interfering substances behind. The assay is based on the specific binding of copper ions to protein.

  • Works in the presence of 2% SDS, 1% DTT, 8M urea, 2M thiourea, 4% CHAPS, 2% Pharmalyte™, and 2% IPG Buffer
  • Quantitatively precipitates proteins while leaving interfering substances behind
  • Linear response in the range of 0 to 50μg protein, with sample volumes of 1 to 50μL
  • Assay time: one hour

2-D electrophoresis, SDS-PAGE, IEF

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) 2D Electrophoresis, SDS-PAGE, IEF
Includes 250mL Precipitant, 250mL Co Precipitant, 50mL Copper Solution, 2 x 250mL Color reagent A, 5mL Color reagent B, 5mL BSA standard (2mg/mL)
Sample Volume 1 to 50μL
Content And Storage 4° to 8°C
Product Type 2D Quant kit
Quantity 500 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.