missing translation for 'onlineSavingsMsg'
Learn More

2-Debenzoyl Paclitaxel 2-Pentanoate, TRC

Product Code. 30135270
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135270 2.5 mg 2.50mg
30134580 25 mg 25mg
30006922 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135270 Supplier Toronto Research Chemicals Supplier No. TRCD2045002.5MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213767-22-7
Molecular Formula C45 H55 N O14
Molecular Weight (g/mol) 833.92
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-12-[(1-oxopentyl)oxy]-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-
IUPAC Name [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4,12-diacetyloxy-15-[(2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoyl]oxy-1,9-dihydroxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] pentanoate
SMILES CCCCC(=O)O[C@H]1[C@@H]2[C@@]3(CO[C@@H]3C[C@H](O)[C@@]2(C)C(=O)[C@H](OC(=O)C)C4=C(C)[C@H](C[C@]1(O)C4(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)c5ccccc5)c6ccccc6)OC(=O)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C45H55NO14/c1-8-9-20-32(50)59-39-37-43(7,30(49)21-31-44(37,23-56-31)60-26(4)48)38(52)36(57-25(3)47)33-24(2)29(22-45(39,55)42(33,5)6)58-41(54)35(51)34(27-16-12-10-13-17-27)46-40(53)28-18-14-11-15-19-28/h10-19,29-31,34-37,39,49,51,55H,8-9,20-23H2,1-7H3,(H,46,53)/t29-,30-,31+,34-,35+,36+,37-,39-,43+,44-,45+/m0/s1
Formula Weight 833.3623
Chemical Name or Material 2-Debenzoyl Paclitaxel 2-Pentanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.