missing translation for 'onlineSavingsMsg'
Learn More

2-Decanol, 98%

Product Code. 10642572 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10642572 5 g 5g
10623142 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10642572 Supplier Thermo Scientific Acros Supplier No. 331600050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1120-06-5 | C10H22O | 158.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1120-06-5
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
MDL Number MFCD00004594
InChI Key ACUZDYFTRHEKOS-UHFFFAOYSA-N
Synonym 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
PubChem CID 14254
IUPAC Name decan-2-ol
SMILES CCCCCCCCC(C)O

Spezifikation

CAS Min % 97.5
CAS Max % 100.0
Melting Point -5°C
Color Colorless
Density 0.8200g/mL
Boiling Point 211°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CH(OH)CH3
Refractive Index 1.4330 to 1.435
Quantity 5 g
Beilstein 01, I, 213
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble in water
Formula Weight 158.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Decanol, 95%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.