missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-2-fluoro-D-glucose, TRC

Product Code. 30136160
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136160 25 mg 25mg
30134696 100 mg 100mg
17931045 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136160 Supplier Toronto Research Chemicals Supplier No. TRCD23450025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 29702-43-0
Molecular Formula C6 H11 F O5
Molecular Weight (g/mol) 182.15
Synonym 2-Deoxy-2-fluoro-D-glucose, 2-Deoxy-2-fluoro-D-glucopyranose, 2-Deoxy-2-fluoroglucose, 2-Fluoro-2-deoxy-D-glucose, FDG, Fludeoxyglucose
IUPAC Name (2R,3S,4R,5R)-2-fluoro-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](F)C=O

Especificaciones

Quantity 25 mg
InChI Formula InChI=1S/C6H11FO5/c7-3(1-8)5(11)6(12)4(10)2-9/h1,3-6,9-12H,2H2/t3-,4+,5+,6+/m0/s1
Formula Weight 182.0591
Percent Purity >95%
Chemical Name or Material 2-Deoxy-2-fluoro-D-glucose
Grade HPLC
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.