missing translation for 'onlineSavingsMsg'
Learn More

2-deoxy-D-galactopyranose, MP Biomedicals™

Product Code. 11452202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452202 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11452202 Supplier MP Biomedicals Supplier No. 0215080005
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    310.80 € / 5g
    Save 17.20 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 0.996
Quantity 5 g
IUPAC Name (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
Formula Weight 164.2
Percent Purity 99.6%
Grade Research
Physical Form Powder
Chemical Name or Material 2-Deoxy-D-galactopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.