missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-glucose, 98%

Product Code. 11321867
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11321867 5 g 5g
11311867 1 g 1g
11476740 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11321867 Supplier Thermo Scientific Chemicals Supplier No. L07338.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 154-17-6 | C6H12O5 | 164.16 g/mol

2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-17-6
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151328
InChI Key PMMURAAUARKVCB-PHUJZJCSNA-N
Synonym deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
PubChem CID 17751002
IUPAC Name (3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O

Specifications

Melting Point 147°C to 152°C
Quantity 5 g
Beilstein 1723331
Merck Index 14,2904
Solubility Information Soluble in water.
Optical Rotation +47° (c=0.5 in Water, 45h)
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 2-Deoxy-D-glucose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.