missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-N-(3-oxo-1-propenyl)adenosine, TRC

Product Code. 30359882
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359882 10 mg 10mg
30368200 5 mg 5mg
30352253 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359882 Supplier Toronto Research Chemicals Supplier No. TRCD33005010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178427-43-5
Molecular Formula C13H15N5O4
Molecular Weight (g/mol) 305.29
Synonym 2'-Deoxy-N-(3-oxo-1-propen-1-yl)adenosine, 3-(2'-Deoxyribofuranosyl-N6-adenosinyl)propenal
SMILES OC[C@H]1O[C@H](C[C@@H]1O)n2cnc3c(N\C=C\C=O)ncnc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H15N5O4/c19-3-1-2-14-12-11-13(16-6-15-12)18(7-17-11)10-4-8(21)9(5-20)22-10/h1-3,6-10,20-21H,4-5H2,(H,14,15,16)/b2-1+/t8-,9+,10+/m0/s1
Formula Weight 305.1124
Chemical Name or Material 2'-Deoxy-N-(3-oxo-1-propenyl)adenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.