missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyadenosine hydrate, 99%

Product Code. 10265770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265770 1 g Glass bottle 1g
10081830 5 g Glass bottle 5g
10771355 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265770 Supplier Thermo Scientific Chemicals Supplier No. 154990010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16373-93-6 | C10H13N5O3 | 251.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16373-93-6
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00149364
InChI Key OLXZPDWKRNYJJZ-YUZWJPFSNA-N
Synonym 2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate
PubChem CID 9549172
SMILES NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 185.0°C to 187.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: 0.3% in water (20°C). Other solubilities: soluble in dmf,formamide,hot dioxane,methanol,ethanol,insoluble in hydrocarbons
IUPAC Name (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol;hydrate
Formula Weight 251.24
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxyadenosine hydrate
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Deoxyadenosine hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.