missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 2'-Deoxyadenosine monohydrate

Product Code. 16499346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499346 Supplier MedChemExpress Supplier No. HYW011683

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2'-Deoxyadenosine monohydrate is a deoxyribonucleoside. A building block in the chemical synthesis.

Specifications

Molecular Weight (g/mol) 269.26 g/mol
Product Type Natural Products
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Shipping Condition Room temperature
Purity 95.66%
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (371.39 mM∣Need ultrasonic)
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 16373-93-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.