missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyadenosine monohydrate, 99%

Product Code. 11429103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429103 1 g 1g
11439103 5 g 5g
11449103 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429103 Supplier Thermo Scientific Chemicals Supplier No. A11166.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16373-93-6 | C10H13N5O3 | 251.25 g/mol

2?-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2?-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16373-93-6
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00149364
InChI Key OLXZPDWKRNYJJZ-YUZWJPFSNA-N
Synonym 2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate
PubChem CID 9549172
IUPAC Name (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol;hydrate
SMILES NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

Melting Point 186°C to 189°C
Quantity 1 g
Beilstein 5191174
Solubility Information Soluble in water.
Optical Rotation −25° (c=0.5 water)
Formula Weight 269.26 (251.24 Anhydrous)
Percent Purity 99%
Chemical Name or Material 2'-Deoxyadenosine monohydrate
compliance-icons
Product Identifier
  • 2-Deoxyadenosine monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2'-Deoxyadenosine hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.