missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine Monohydrate, TRC

Product Code. 30136422
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136422 500 mg 500mg
30004381 250 mg 250mg
30004739 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136422 Supplier Toronto Research Chemicals Supplier No. TRCD231620500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16373-93-6
Molecular Formula C10 H13 N5 O3 . H2 O
Molecular Weight (g/mol) 269.26
Synonym 2'-Deoxyadenosine Hydrate (1:1), Adenosine, 2'-deoxy-, monohydrate (8CI,9CI), 2'-Deoxyadenosine monohydrate, Deoxyadenosine monohydrate
IUPAC Name (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol;hydrate
SMILES O.Nc1ncnc2c1ncn2[C@H]3C[C@H](O)[C@@H](CO)O3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H13N5O3.H2O/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(17)6(2-16)18-7;/h3-7,16-17H,1-2H2,(H2,11,12,13);1H2/t5-,6+,7+;/m0./s1
Formula Weight 269.1124
Chemical Name or Material 2'-Deoxyadenosine Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.