missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine

Product Code. p-7042590
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475329 1 g 1g
15485329 5 g 5g
15495329 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475329 Supplier Thermo Scientific Chemicals Supplier No. J63062.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 951-77-9 | C9H13N3O4 | 227.22 g/mol

2?-Deoxycytidine (deoxyC) is one of the deoxynucleosides which after phosphorylation to dCTP is used to synthesis DNA via various DNA polymerases or reverse transcriptases. 2?-Deoxycytidine (deoxyC) is the substrate for deoxycytidine deaminase (EC 3.5.4.14) which converts it into 2?-deoxyuridine. 2?-Deoxycytidine is phosphorylated to the nucleotide dCMP by the enzyme deoxycytidine kinase (DCK).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxycytidine (deoxyC) is one of the deoxynucleosides which after phosphorylation to dCTP is used to synthesis DNA via various DNA polymerases or reverse transcriptases. 2′-Deoxycytidine (deoxyC) is the substrate for deoxycytidine deaminase (EC 3.5.4.14) which converts it into 2′-deoxyuridine. 2′-Deoxycytidine is phosphorylated to the nucleotide dCMP by the enzyme deoxycytidine kinase (DCK).

Solubility
Soluble in water, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-77-9
Molecular Formula C9H13N3O4
Molecular Weight (g/mol) 227.22
MDL Number MFCD00006547
InChI Key CKTSBUTUHBMZGZ-JORGKRSHNA-N
Synonym 2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
PubChem CID 13711
ChEBI CHEBI:15698
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Melting Point 209°C to 211°C
Color White
Quantity 1 g
Beilstein 87567
Solubility Information Soluble in water,DMSO.
Formula Weight 227.22
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxycytidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.