missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine hydrochloride, 98%

Product Code. 15520574 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15520574 25 g 25g
15500574 1 g 1g
15510574 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15520574 Fornitore Thermo Scientific Acros N. del fornitore 458510250
 Restricted Item
Estimated Shipment: 09-11-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 3992-42-5 | C9H14ClN3O4 | 263.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3992-42-5
Molecular Formula C9H14ClN3O4
Molecular Weight (g/mol) 263.68
MDL Number MFCD00012840
InChI Key LTKCXZGFJFAPLY-VOBHXVAMNA-N
Synonym 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl
PubChem CID 107488
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifica

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 263.68
Percent Purity 98%
Chemical Name or Material 2'-Deoxycytidine hydrochloride

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.