missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine hydrochloride, 99%

Product Code. 11399005
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399005 1 g 1g
11309015 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399005 Supplier Thermo Scientific Chemicals Supplier No. L14153.03
 Restricted Item
Estimated Shipment: 25-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3992-42-5 | C9H14ClN3O4 | 263.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3992-42-5
Molecular Formula C9H14ClN3O4
Molecular Weight (g/mol) 263.68
MDL Number MFCD00012840
InChI Key LTKCXZGFJFAPLY-VOBHXVAMNA-N
Synonym 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl
PubChem CID 107488
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Melting Point ∽170°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 3576124
Sensitivity Hygroscopic
Optical Rotation +57° (c=1 in Water)
Formula Weight 263.68
Percent Purity 99%
Chemical Name or Material 2'-Deoxycytidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.