missing translation for 'onlineSavingsMsg'
Learn More

2'-deoxycytidine hydrochloride, MP Biomedicals™

Product Code. 11463450 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463450 10 g 10g
11443450 1 g 1g
11453450 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463450 Supplier MP Biomedicals Supplier No. 0210148310
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3992-42-5
Molecular Formula C9H14ClN3O4
Molecular Weight (g/mol) 263.68
MDL Number MFCD00012840
InChI Key LTKCXZGFJFAPLY-VOBHXVAMNA-N
Synonym 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl
PubChem CID 107488
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Color White
Assay Percent Range 99.9%
Quantity 10 g
Formula Weight 263.68
Physical Form Powder
Chemical Name or Material 2'-Deoxycytidine hydrochloride, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.