missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyguanosine hydrate, 99+%, Thermo Scientific Chemicals

Product Code. 10710575 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
5 g
Unit Size:
100mg
1g
25mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10710575 1 g 1g
10348510 25 mg 25mg
10216240 100 mg 100mg
10464905 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10710575 Supplier Thermo Scientific Acros Supplier No. 226730010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 961-07-9 | C10H13N5O4 | 267.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 961-07-9
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00150760
InChI Key YKBGVTZYEHREMT-DWFKHXNSNA-N
Synonym 2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl
PubChem CID 187790
ChEBI CHEBI:17172
IUPAC Name 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1

Specifications

Absorbance 1.12 to 1.18 (A250/A260) 0.65 to 0.69 (A280/A260)
CAS Min % 99.0
CAS Max % 100.0
Melting Point 300°C
Color White
Elemental Analysis (on dry substance) :
Flash Point 300°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Spectrophotometry 98% min. (lambda max. 252.5nm,pH 7) molar absorptivity 13700+/-4%,on dry basis
Formula Weight 267.12
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 2′-Deoxyguanosine hydrate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.