missing translation for 'onlineSavingsMsg'
Learn More

2’-Deoxyguanosine Monohydrate, TRC

Product Code. 30361951
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361951 25 mg 25mg
30355057 100 mg 100mg
30355446 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361951 Supplier Toronto Research Chemicals Supplier No. TRCD23955025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312693-72-4
Molecular Formula C10 H13 N5 O4 . H2 O
Molecular Weight (g/mol) 285.26
Synonym 2'-Deoxyguanosine Hydrate (1:1), Guanosine, 2'-deoxy-, monohydrate (9CI), 2'-Deoxyguanosine monohydrate
IUPAC Name 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one;hydrate
SMILES O.NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13N5O4.H2O/c11-10-13-8-7(9(18)14-10)12-3-15(8)6-1-4(17)5(2-16)19-6;/h3-6,16-17H,1-2H2,(H3,11,13,14,18);1H2/t4-,5+,6+;/m0./s1
Formula Weight 285.1073
Chemical Name or Material 2'-Deoxyguanosine Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.