missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyinosine, 98+%, Thermo Scientific™

Product Code. 10723421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723421 100 mg Glass bottle 100mg
10166823 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723421 Supplier Thermo Scientific Acros Supplier No. 226361000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 890-38-0
Molecular Formula C10H12N4O4
Molecular Weight (g/mol) 252.23
MDL Number MFCD00005762
InChI Key VGONTNSXDCQUGY-YUZWJPFSNA-N
Synonym 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine
PubChem CID 65058
ChEBI CHEBI:28997
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point >250.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Specific Rotation + 21.00
IUPAC Name 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Formula Weight 252.23
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 2'-Deoxyinosine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.