missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyuridine-5'-triphosphate trisodium salt, 95%

Product Code. 15420387
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420387 100 mg 100mg
15410387 25 mg 25mg
15430387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420387 Supplier Thermo Scientific Alfa Aesar Supplier No. J64716.MC
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102814-08-4 | C9H15N2Na3O14P3 | 537.11 g/mol

2'-Deoxyuridine-5'-triphosphate disodium salt is used in conjunction with uracil DNA glycosylase to prevent carryover contamination in PCR. The dUTP is substituted for dTTP in the PCR reaction, used to study the distribution, specificity and kinetics of deoxyuridine 5?-triphosphate nucleotidohydrolase(s) (dUTPase, EC 3.6.1.23). dUTPase are important enzymes that protect cells from the incorporation of dUTP into DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyuridine-5′-triphosphate disodium salt is used in conjunction with uracil DNA glycosylase to prevent carryover contamination in PCR. The dUTP is substituted for dTTP in the PCR reaction, used to study the distribution, specificity and kinetics of deoxyuridine 5′-triphosphate nucleotidohydrolase(s) (dUTPase, EC 3.6.1.23). dUTPase are important enzymes that protect cells from the incorporation of dUTP into DNA.

Solubility
Soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102814-08-4
Molecular Formula C9H15N2Na3O14P3
Molecular Weight (g/mol) 537.11
MDL Number MFCD00084701
InChI Key AYFBRVJQNMGUSV-MILVPLDLSA-N
Synonym 2'-deoxyuridine-5'-triphosphate trisodium salt
PubChem CID 131843218
IUPAC Name [[(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O.[Na].[Na].[Na]

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water.
Formula Weight 537.11
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2'-Deoxyuridine-5'-triphosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.