missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyuridine, MedChemExpress

Product Code. 16478886
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478886 Supplier Medchem Express Supplier No. HYD0186
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2'-Deoxyuridine could increase chromosome breakage and results in a decreased thymidylate synthetase activity. A known use of 2'-Deoxyuridine is as a precursor in the synthesis of Edoxudine.

2'-Deoxyuridine could increase chromosome breakage and results in a decreased thymidylate synthetase activity. A known use of 2'-Deoxyuridine is as a precursor in the synthesis of Edoxudine.

Specifications

Chemical Name or Material 2'-Deoxyuridine
CAS 951-78-0
Color White
Purity Grade Notes Research
Molecular Formula C9H12N2O5
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (438.21 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C=C2)=O)=O)O1)O
Molecular Weight (g/mol) 228.2
Formula Weight 228.2
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.