missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethyl 3-amino-4-(2-(diethylamino)ethoxy)benzoate Hydrochloride, TRC

Product Code. 30350027
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350027 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350027 Supplier Toronto Research Chemicals Supplier No. TRCD36702010MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Diethylamino)ethyl 3-amino-4-(2-(diethylamino)ethoxy)benzoate Hydrochloride
CAS 2518227-82-0
Molecular Formula C19H33N3O3 • (HCl)
Quantity 10 mg
Synonym Proparacaine Hydrochloride Impurity
InChI Formula InChI=1S/C19H33N3O3.ClH/c1-5-21(6-2)11-13-24-18-10-9-16(15-17(18)20)19(23)25-14-12-22(7-3)8-4;/h9-10,15H,5-8,11-14,20H2,1-4H3;1H
SMILES NC1=CC(C(OCCN(CC)CC)=O)=CC=C1OCCN(CC)CC.Cl
IUPAC Name 2-(diethylamino)ethyl 3-amino-4-(2-(diethylamino)ethoxy)benzoate hydrochloride
Molecular Weight (g/mol) 387.94
Formula Weight 387.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.