missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Diisopropylphosphino)ethylamine, 10 wt.% solution in THF, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 mL
Packaging:
Glass bottle
Unit Size:
2.5mL
Specifications
Specifications
| CAS | 1053657-14-9, 109-99-9 |
| Color | White |
| Flash Point | −21°C |
| Packaging | Glass bottle |
| Molecular Formula | C8H20NP |
| Linear Formula | ((CH3)2CH)2PCH2CH2NH2 |
| MDL Number | MFCD11044863 |
| Quantity | 2.5 mL |
| Synonym | 2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity |
| InChI Key | QUVUVMRUSAKOPB-UHFFFAOYSA-N |
| Show More |
Safety and Handling
Product Identifier
- 2-(Diisopropylphosphino)ethylamine, 10wt.% solution in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction