missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)-3-quinolinecarbaldehyde, 97+%, Thermo Scientific™

Product Code. 10299212
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10299212 250 mg 250mg
10209222 1 g 1g
10507474 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10299212 Supplier Thermo Scientific Maybridge Supplier No. MO07114CB
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 728035-61-8
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD06010203
InChI Key RCRLBHNGTPRGJQ-UHFFFAOYSA-N
PubChem CID 1076116
IUPAC Name 2-(dimethylamino)quinoline-3-carbaldehyde
SMILES CN(C)C1=C(C=O)C=C2C=CC=CC2=N1

Specifications

Assay Percent Range 97%
Quantity 250 mg
Synonym 2-dimethylamino quinoline-3-carbaldehyde, 2-dimethylamino-quinoline-3-carbaldehyde, 2-dimethylamino-3-quinolinecarbaldehyde, 3-quinolinecarboxaldehyde,2-dimethylamino, 2-dimethylamino-3-quinoline carbaldehyde, 2-dimethylamino quinoline-3-carboxaldehyde, 3-quinolinecarboxaldehyde, 2-dimethylamino
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)-3-quinolinecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.