missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethanol, 99+%

Product Code. 11485647
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
This item is not returnable. View return policy

Product Code. 11485647

Brand: Thermo Scientific Alfa Aesar B23616.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-01-0 | C4H11NO | 89.138 g/mol

2-(Dimethylamino)ethanol is used as corrosion inhibitor, anti-scaling agent, paint additive, coating additive and solids separation agent. It is also used as an intermediate for active pharmaceutical ingredients and dyes. It serves as a curing agent for polyurethanes and epoxy resins. Further, it is used as an additive to boiler water. In addition to this, it is used therapeutically as a CNS stimulant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, alcohol, ether, acetone and benzene.

Notes
Incompatible with strong oxidizers and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-01-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002846
InChI Key UEEJHVSXFDXPFK-UHFFFAOYSA-N
Synonym 2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon
PubChem CID 7902
ChEBI CHEBI:271436
IUPAC Name 2-(dimethylamino)ethanol
SMILES CN(C)CCO

Specifications

Melting Point -60°C
Density 0.886
Boiling Point 134°C to 136°C
Flash Point 40°C (104°F)
Odor Ammonia-like
Linear Formula (CH3)2NCH2CH2OH
Refractive Index 1.4295
Quantity 250 mL
UN Number UN2051
Beilstein 1209235
Sensitivity Hygroscopic
Merck Index 14,2843
Solubility Information Miscible with water,alcohol,ether,acetone and benzene.
Formula Weight 89.14
Percent Purity ≥99%
Chemical Name or Material 2-(Dimethylamino)ethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-(Dimethylamino)ethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-(Dimethylamino)ethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.