missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol

Product Code. 11333117
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333117 500 mL 500mL
11323117 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333117 Supplier Thermo Scientific Alfa Aesar Supplier No. L13507.AP
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2867-47-2 | C8H15NO2 | 157.213 g/mol

2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.

Solubility
Soluble in water. (1.000 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture and light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2867-47-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00008589
InChI Key JKNCOURZONDCGV-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl methacrylate, dimethylaminoethyl methacrylate, ageflex fm-1, dmaema, usaf rh-3, 2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester, 2-n,n-dimethylamino ethyl methacrylate, n,n-dimethylaminoethyl methacrylate, 2-dimethylaminoethyl methacrylate, n,n-dimethylethanolamine methacrylate
PubChem CID 17869
IUPAC Name 2-(dimethylamino)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN(C)C

Specifications

Melting Point ∼-30°C
Density 0.932
Boiling Point 186°C to 188°C
Flash Point 70°C (158°F)
Refractive Index 1.44
Quantity 500 mL
UN Number UN2522
Beilstein 1757048
Solubility Information Soluble in water. (1.000g/L at 20°C).
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)ethyl methacrylate, stabilized with ≈0.2% 4-methoxyphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Dimethylamino)ethyl methacrylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Dimethylaminoethyl methacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.