missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl Methacrylate (Stabilized with 0.2% 4-methoxyphenol), TRC

Product Code. 30136229
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136229 5 g 5g
30134699 10 g 10g
30009911 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30136229 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD4617055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2867-47-2
Formula Weight 157.1103
InChI Formula InChI=1S/C8H15NO2/c1-7(2)8(10)11-6-5-9(3)4/h1,5-6H2,2-4H3
IUPAC Name 2-(dimethylamino)ethyl 2-methylprop-2-enoate
Molecular Formula C8 H15 N O2
Molecular Weight (g/mol) 157.21
SMILES CN(C)CCOC(=O)C(=C)C
Synonym 2-(Dimethylamino)ethyl Methacrylate, 2-Propenoic acid, 2-methyl-, 2-(dimethylamino)ethyl ester, Methacrylic acid, 2-(dimethylamino)ethyl ester (6CI,8CI), (2-Methacryloyloxyethyl)dimethylamine, (Dimethylamino)ethyl methacrylate, 2-(N,N-Dimethylamino)ethyl methacrylate, Acryester DM, Ageflex FA 1, Ageflex FM 1, Ageflex FM 2, DM, DM (methacrylate), DMA 200, DMAEMA, Dimethyl[2-(methacryloyloxy)ethyl]amine, GE 720, Light Ester DM, MADAM, Methacryl Ester DM, Methacrylate DMA, Methacryloyloxyethyldimethylamine, Mhoromer BM 601, N,N-Dimethylaminoethyl methacrylate, N,N-Dimethylethanolamine methacrylate, N,N'-Dimethylamino-2-ethyl methacrylate, NSC 20959, Pentanoic acid, 5-(dimethylamino)-2-methylene-, β-(N,N-Dimethylamino)ethyl methacrylate, β-Dimethylaminoethyl methacrylate
Chemical Name or Material 2-(Dimethylamino)ethyl Methacrylate

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.