missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoisopropyl Chloride Hydrochloride, TRC

Product Code. 30388430
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388430 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388430 Supplier Toronto Research Chemicals Supplier No. TRCD471060100G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Dimethylaminoisopropyl Chloride Hydrochloride
CAS 4584-49-0
Molecular Formula C5H13Cl2N
Quantity 100 g
Synonym 2-Chloro-N,N-dimethyl-1-propanamine Hydrochloride, 2-Chloro-N,N-dimethyl-propylamine Hydrochloride, (2-Chloropropyl)dimethylamine Hydrochloride, (2-Chloropropyl)dimethylamine Monohydrochloride, 1-(Dimethylamino)-2-chloropropane Hydrochloride, 1-Dimethylamino-2-propyl Chloride Hydrochloride, 1-Dimethylaminopropan-2-yl Chloride Hydrochloride, 1-Methyl-2-dimethylaminoethyl Chloride Hydrochloride, 2-(Dimethylamino)-1-methylethyl Chloride Hydrochloride, 2-(Dimethylamino)isopropyl Chloride Hydrochloride, 2-Chloro-N,N-dimethyl-1-propanamine Hydrochloride, 2-Chloro-N,N-dimethylaminopropane Hydrochloride, 2-Chloro-N',N'-dimethylaminopropane Hydrochloride, Dimethylaminoisopropyl Chloride Hydrochloride, N,N-Dimethyl-2-chloropropylamine Hydrochloride, NSC 53532, NSC 5367
InChI Formula InChI=1S/C5H12ClN.ClH/c1-5(6)4-7(2)3;/h5H,4H2,1-3H3;1H
SMILES Cl.CC(Cl)CN(C)C
IUPAC Name 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 158.07
Formula Weight 157.0425
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.