missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminopyridine, 98%

Product Code. 11906301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11906301 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11906301 Supplier Thermo Scientific Acros Supplier No. 146680250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5683-33-0 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5683-33-0
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9840g/mL
Boiling Point 191.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C7H10N2
Refractive Index 1.559
MDL Number MFCD00006261
Quantity 25 g
UN Number 1992
Beilstein 22,I,629
Specific Gravity 0.984
Synonym 2-dimethylaminopyridine, 2-dimethylaminopyridin, dimethylaminopyridine, 2-dimethylamino pyridine, 2-dimethylaminopyridine cobalt complex, 2-pyridinamine, n,n-dimethyl, dimethylamino-2 pyridine, dimethylaminopyridin, unii-18q95u3z8h, pyridine, 2-dimethylamino
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in diluted acids
InChI Key PSHKMPUSSFXUIA-UHFFFAOYSA-N
SMILES CN(C)C1=CC=CC=N1
IUPAC Name N,N-dimethylpyridin-2-amine
Molecular Weight (g/mol) 122.17
PubChem CID 21885
Formula Weight 122.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Dimethylaminopyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Dimethylaminopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.