missing translation for 'onlineSavingsMsg'
Learn More

2-Ethenyl-3,4-dihydroquinazolin-4-one, TRC

Product Code. 30138001
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138001 50 mg 50mg
30136409 250 mg 250mg
30010500 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138001 Supplier Toronto Research Chemicals Supplier No. TRCB12635050MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91634-12-7
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.18
IUPAC Name 2-ethenyl-3H-quinazolin-4-one
SMILES Oc1nc(C=C)nc2ccccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H8N2O/c1-2-9-11-8-6-4-3-5-7(8)10(13)12-9/h2-6H,1H2,(H,11,12,13)
Formula Weight 172.0637
Chemical Name or Material 2-Ethenyl-3,4-dihydroquinazolin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.