missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, 99%

Product Code. 11454734
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454734 10 g 10g
11464734 50 g 50g
11474734 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454734 Supplier Thermo Scientific Alfa Aesar Supplier No. A13724.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16357-59-8 | C14H17NO3 | 247.29 g/mol

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

Solubility
Soluble in 95% Ethanol (50 mg/ml). Insoluble in water.

Notes
Moisture Sensitive. Store at 4°C. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16357-59-8
Molecular Formula C14H17NO3
Molecular Weight (g/mol) 247.29
MDL Number MFCD00006703
InChI Key GKQLYSROISKDLL-UHFFFAOYNA-N
Synonym eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
PubChem CID 27833
IUPAC Name ethyl 2-ethoxy-2H-quinoline-1-carboxylate
SMILES CCOC1C=CC2=CC=CC=C2N1C(=O)OCC

Specifications

Melting Point 63°C to 66°C
Boiling Point 125°C to 128°C (0.1 mmHg)
Quantity 10 g
Beilstein 533048
Sensitivity Moisture sensitive
Merck Index 14,3518
Solubility Information Soluble in 95% Ethanol (50mg/ml). Insoluble in water.
Formula Weight 247.29
Percent Purity 99%
Chemical Name or Material 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.