missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzenesulfonyl chloride, 96%

Product Code. 15417267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417267 5 g 5g
15407267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417267 Supplier Thermo Scientific Alfa Aesar Supplier No. H34325.06
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 68800-33-9 | C8H9ClO3S | 220.667 g/mol

2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68800-33-9
Molecular Formula C8H9ClO3S
Molecular Weight (g/mol) 220.667
MDL Number MFCD08706420
InChI Key RWURYJNJYODVAL-UHFFFAOYSA-N
Synonym 2-ethoxy-benzenesulfonyl chloride, 2-ethoxybenzene-1-sulfonyl chloride, 2-ethoxyphenylsulfonyl chloride, benzenesulfonylchloride, 2-ethoxy
PubChem CID 18944993
IUPAC Name 2-ethoxybenzenesulfonyl chloride
SMILES CCOC1=CC=CC=C1S(=O)(=O)Cl

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 220.67
Percent Purity 96%
Chemical Name or Material 2-Ethoxybenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.