missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)ethylzinc bromide, 0.5M in THF

Product Code. 15413537
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413537 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413537 Supplier Thermo Scientific Alfa Aesar Supplier No. H26739.AD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 193065-68-8 | C5H10BrO2Zn- | 247.417 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In many of the reactions, organozinc compounds appear as intermediates. Some of the synthetic uses of organozinc compounds are: for the stereo selective alkylation reaction of cyclic ketones, conjugate addition reaction to alpha, be
TRUSTED_SUSTAINABILITY

Specifications

CAS 193065-68-8
Density 0.972
Flash Point −17°C (1°F)
Molecular Formula C5H10BrO2Zn-
MDL Number MFCD00671976
Quantity 50 mL
UN Number UN3399
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 2-ethoxycarbonyl ethylzinc bromide, 0.5m in thf
Solubility Information Reacts with water.
InChI Key APUPJTIEDQLKSC-UHFFFAOYSA-M
SMILES CCC(=O)OCC.[Zn].[Br-]
IUPAC Name ethyl propanoate;zinc;bromide
Molecular Weight (g/mol) 247.417
PubChem CID 131668631
Formula Weight 246.42
Chemical Name or Material 2-(Ethoxycarbonyl)ethylzinc bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.