missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)phenyl isothiocyanate, 98+%, Thermo Scientific™

Product Code. 11383236
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383236 5 g 5g
11373236 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383236 Supplier Thermo Scientific Alfa Aesar Supplier No. L12569.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99960-09-5
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.247
MDL Number MFCD00041063
InChI Key TYEPWLJHQLYWSR-UHFFFAOYSA-N
Synonym 2-ethoxycarbonylphenyl isothiocyanate, 2-ethoxycarbonyl phenyl isothiocyanate, 2-isothiocyanatobenzoic acid ethyl ester, 2-isothiocyanato-benzoic acid ethyl ester, acmc-20aon5, #, 2-ethoxycarbonylphenylisothiocyanate, ethyl 2-isothiocyanatobenzoate, isothiocyanatobenzoic acid ethyl ester, benzoic acid,2-isothiocyanato-,ethyl ester
PubChem CID 523235
IUPAC Name ethyl 2-isothiocyanatobenzoate
SMILES CCOC(=O)C1=CC=CC=C1N=C=S

Specifications

Density 1.201
Boiling Point 144°C to 145°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.616
Quantity 5 g
UN Number UN2922
Beilstein 3267738
Sensitivity Moisture sensitive
Formula Weight 207.25
Percent Purity ≥98%
Chemical Name or Material 2-(Ethoxycarbonyl)phenyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.