missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxycarbonylphenylboronic acid, TRC

Product Code. 30383336
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30383336 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30383336

Brand: Toronto Research Chemicals TRCE895025500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethoxycarbonylphenylboronic acid
CAS 380430-53-5
Molecular Formula C9H11BO4
Quantity 500 mg
InChI Formula InChI=1S/C9H11BO4/c1-2-14-9(11)7-5-3-4-6-8(7)10(12)13/h3-6,12-13H,2H2,1H3
SMILES CCOC(=O)c1ccccc1B(O)O
IUPAC Name (2-ethoxycarbonylphenyl)boronic acid
Molecular Weight (g/mol) 193.99
Formula Weight 194.075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.